Skip to Content
Merck
All Photos(1)

Key Documents

T-069

Supelco

Tilidine-D6 hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17D6H17NO2 · HCl
Molecular Weight:
315.87
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CCOC([C@]1(C2=CC=CC=C2)CCC=C[C@H]1N(C([2H])([2H])[2H])C([2H])([2H])[2H])=O.Cl

InChI

1S/C17H23NO2.ClH/c1-4-20-16(19)17(14-10-6-5-7-11-14)13-9-8-12-15(17)18(2)3;/h5-8,10-12,15H,4,9,13H2,1-3H3;1H/t15-,17+;/m1./s1/i2D3,3D3;

InChI key

MUWDJVKYGSDUSH-YGQHATDOSA-N

General description

This stable-labeled internal standard is suitable for quantification of tilidine levels in GC/MS or LC/MS testing methods for clinical toxicology, forensic analysis, urine drug testing, or isotope dilution applications. Tilidine is a synthetic opioid analgesic used mainly in Germany, Switzerland and Belgium for treatment of moderate to severe pain, both acute and chronic. While tilidine is considered a moderately-potent opioid, its active metabolite nortilidine possesses analgesic effects roughly equivalent to that of morphine.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service