Skip to Content
Merck
All Photos(1)

Key Documents

N-136

Supelco

Norscopolamine solution

1 mg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19NO4
CAS Number:
Molecular Weight:
289.33
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in acetonitrile

format

single component solution

storage temp.

−20°C

SMILES string

O=C(O[C@@H]1C[C@H](N2[H])[C@H](O3)[C@H]3[C@H]2C1)[C@H](CO)C4=CC=CC=C4

InChI

1S/C16H19NO4/c18-8-11(9-4-2-1-3-5-9)16(19)20-10-6-12-14-15(21-14)13(7-10)17-12/h1-5,10-15,17-18H,6-8H2/t10?,11-,12-,13+,14-,15+/m1/s1

InChI key

MOYZEMOPQDTDHA-YQYZCKNZSA-N

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service