Skip to Content
Merck
All Photos(1)

Key Documents

N-084

Supelco

Norsertraline-13C6 hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€239.00

€239.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€239.00

About This Item

Empirical Formula (Hill Notation):
13C6C10H15NCl2 · HCl
Molecular Weight:
334.62
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€239.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

ClC1=CC(C2[13C]3=[13CH][13CH]=[13CH][13CH]=[13C]3C(N)CC2)=CC=C1Cl.[H]Cl

InChI

1S/C16H15Cl2N.ClH/c17-14-7-5-10(9-15(14)18)11-6-8-16(19)13-4-2-1-3-12(11)13;/h1-5,7,9,11,16H,6,8,19H2;1H/i1+1,2+1,3+1,4+1,12+1,13+1;

InChI key

YKXHIERZIRLOLD-JQQCCREHSA-N

General description

Norsertraline is a major urinary metabolite and N-desmethyl analog of the SSRI antidepressant sertraline. Marketed under trade names such as Zoloft® and Lustral, sertraline is used to treat major depressive disorder in adults as well as obsessive-compulsive, panic, and social anxiety disorders in both adults and children. Suitable uses for this certified solution standard include as an internal standard for quantitation of norsertraline levels in urine or isotope dilution methods by LC/MS or GC/MS for clinical toxicology, urine drug testing, or forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Zoloft is a registered trademark of Pfizer, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service