Skip to Content
Merck
All Photos(1)

Documents

N-083

Supelco

Normeperidine-D4 solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14D4H15NO2
CAS Number:
Molecular Weight:
237.33
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C1([2H])CNCC([2H])([2H])C1(C(OCC)=O)C2=CC=CC=C2

InChI

1S/C14H19NO2/c1-2-17-13(16)14(8-10-15-11-9-14)12-6-4-3-5-7-12/h3-7,15H,2,8-11H2,1H3/i8D2,9D2

InChI key

QKHMFBKXTNQCTM-LZMSFWOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

An internal standard suitable for normeperidine testing or isotope dilution methods by GC/MS or LC/MS for clinical toxicology, pain prescription monitoring, urine drug testing, or forensic analysis. Normeperidine is the primary urinary metabolite of meperidine, a fast-acting opioid analgesic commonly sold under the trade name Demerol® and indicated for the treatment of moderate to severe pain.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Demerol is a registered trademark of Sanofi-Aventis
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service