Skip to Content
Merck
All Photos(1)

Key Documents

N-030

Supelco

Norfentanyl-D5 oxalate solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16D5H17N2O5
CAS Number:
Molecular Weight:
327.39
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland)

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C1=C([2H])C([2H])=C(N(C2CCNCC2)C(CC)=O)C([2H])=C1[2H].O=C(C(O)=O)O

InChI

1S/C14H20N2O.C2H2O4/c1-2-14(17)16(12-6-4-3-5-7-12)13-8-10-15-11-9-13;3-1(4)2(5)6/h3-7,13,15H,2,8-11H2,1H3;(H,3,4)(H,5,6)/i3D,4D,5D,6D,7D;

InChI key

OLFNESLXXQCODF-BQAHAFBHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

An internal standard suitable for norfentanyl testing of urine, blood, or hair samples or isotope dilution methods by GC/MS or LC/MS in applications such as pain prescription monitoring, clinical toxicology, urine drug testing, or forensic analysis. Norfentanyl is a major urinary metabolite of fentanyl, a potent synthetic analgesic sold under various trade names including Sublimaze, Duragesic®, Actiq®, and Fentora®.

Application

  • Use of Norfentanyl-D5 in forensic toxicology: Norfentanyl-D5 serves as a crucial internal standard in the LC-MS/MS analysis of fentanyl and norfentanyl, particularly in forensic investigations of fatalities involving opioid use(Coopman et al., 2007).

Legal Information

Actiq is a registered trademark of Anesta Corp.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Duragesic is a registered trademark of Johnson & Johnson
Fentora is a registered trademark of Cima Labs, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service