Skip to Content
Merck

Skip To

M-201

Methocarbamol solution

1 mg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mL
Check Cart for Availability
€ 120,00

About This Item

Empirical Formula (Hill Notation):
C11H15NO5
CAS Number:
Molecular Weight:
241.24
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:

€ 120,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

NC(=O)OCC(O)COc1c(cccc1)OC

InChI

1S/C11H15NO5/c1-15-9-4-2-3-5-10(9)16-6-8(13)7-17-11(12)14/h2-5,8,13H,6-7H2,1H3,(H2,12,14)

InChI key

GNXFOGHNGIVQEH-UHFFFAOYSA-N

General description

Methocarbamol, sold under the brand name Robaxin, is a central muscle relaxant used to treat skeletal muscle spasms. This certified solution standard is applicable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from therapeutic drug monitoring to forensic analysis and clinical toxicology.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M-074C-060C-033
grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

format

single component solution

format

single component solution

format

single component solution

format

single component solution

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

concentration

1 mg/mL in acetonitrile

concentration

1.0 mg/mL in methanol

concentration

1.0 mg/mL in methanol (as free base)

concentration

1.0 mg/mL in methanol

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2.0 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters