Skip to Content
Merck
All Photos(1)

Key Documents

M-166

Supelco

5-MeO-MiPT hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22N2O · HCl
CAS Number:
Molecular Weight:
282.81
EC Number:
UNSPSC Code:
41116107
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

InChI

1S/C15H22N2O/c1-11(2)17(3)8-7-12-10-16-15-6-5-13(18-4)9-14(12)15/h5-6,9-11,16H,7-8H2,1-4H3

InChI key

HEDOODBJFVUQMS-UHFFFAOYSA-N

General description

5-MeO-MiPT, known by the street name Moxy or Moxie, is a designer typtamine and analogue of 5-MeO-DiPT that is used as a dissociative and hallucinogen. This certified solution standard is suitable for 5-MeO-MiPT testing methods by GC/MS or LC/MS testing applications in clinical toxicology, urine drug testing, or forensic analysis. The use of 5-MeO-MiPT has recently been reported in Europe.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service