Skip to Content
Merck
All Photos(1)

Key Documents

I-028

Supelco

Interference Mix 6

5-100 μg/mL each component in methanol, Cerilliant®

Synonym(s):

Qualitative Interference Mixture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

form

liquid

composition

Amlodipine besylate, 5 μg/mL (as free base)
Atorvastatin calcium salt, 100 μg/mL (as free base)
Azithromycin dihydrate, 100 μg/mL (as free base)
Bupivacaine hydrochloride monohydrate, 100 μg/mL (as free base)
Cetirizine dihydrochloride, 50 μg/mL (as free base)
Dimenhydrinate, 100 μg/mL
Lisinopril dihydrate, 100 μg/mL (as free base)
Loratadine, 100 μg/mL

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

5-100 μg/mL each component in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

A multi-component solution standard of routinely monitored over-the-counter (OTC) and prescription drugs amlodipine, atorvastatin, cetirizine, azithromycin, bupivacaine, dimenhydrinate, lisinopril, and loratidine. This standard is for qualitative use only in the evaluation of interferences between the mix components and target analytes of interest in a testing laboratory′s analytical methods.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service