Skip to Content
Merck
All Photos(1)

Documents

H-124

Supelco

Hydroxyzine dihydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27ClN2O2 · 2HCl
CAS Number:
Molecular Weight:
447.83
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

OCCOCCN1CCN(C(C2=CC=C(Cl)C=C2)C3=CC=CC=C3)CC1.[H]Cl.[H]Cl

InChI

1S/C21H27ClN2O2.2ClH/c22-20-8-6-19(7-9-20)21(18-4-2-1-3-5-18)24-12-10-23(11-13-24)14-16-26-17-15-25;;/h1-9,21,25H,10-17H2;2*1H

InChI key

ANOMHKZSQFYSBR-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

General description

Hydroxyzine is a commonly prescribed first generation antihistamine with sedative properties. This drug is a common cocaine adulterant along with lidocaine, phenacetin, and levamisole. This Certified Spiking Solution® is suitable for use in a variety of GC/MS or LC/MS testing applications in forensic toxicology, urine drug testing, or clinical chemistry.

Application


  • Antihistamine-Retinoic Acid Receptor Interaction: This publication presents a molecular dynamics simulation and genome-wide meta-analysis to explore the interactions of antihistamines like hydroxyzine dihydrochloride with the retinoic acid receptor beta (RARB), providing insights into possible biochemical pathways and therapeutic potentials (Kim et al., 2023).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service