Skip to Content
Merck
All Photos(1)

Key Documents

F-909

Supelco

Fluoxymesterone solution

1.0 mg/mL in 1,2-dimethoxyethane, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H29FO3
CAS Number:
Molecular Weight:
336.44
UNSPSC Code:
41116107
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in 1,2-dimethoxyethane

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

C[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C

InChI

1S/C20H29FO3/c1-17-8-6-13(22)10-12(17)4-5-15-14-7-9-19(3,24)18(14,2)11-16(23)20(15,17)21/h10,14-16,23-24H,4-9,11H2,1-3H3/t14-,15-,16-,17-,18-,19+,20-/m0/s1

InChI key

YLRFCQOZQXIBAB-HAXSJCRZSA-N

General description

Fluoxymesterone is an anabolic steroid used in the treatment of hypogonadism and delayed puberty. Fluoxymesterone, marketed as Halotestin®, is often used as a performance enhancing drug by athletes. This Certified Spiking Solution® is applicable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from sports testing and urine drug testing to forensic analysis and clinical toxicology.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Halotestin is a registered trademark of Dynamic Sports Nutrition, LLC
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

28.4 °F - closed cup

Flash Point(C)

-2 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cornelia Fürstenberger et al.
Toxicological sciences : an official journal of the Society of Toxicology, 126(2), 353-361 (2012-01-26)
Anabolic androgenic steroids (AAS) are testosterone derivatives used either clinically, in elite sports, or for body shaping with the goal to increase muscle size and strength. Clinically developed compounds and nonclinically tested designer steroids often marketed as food supplements are
Toktam Zohoorian-Abootorabi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 88, 177-191 (2012-01-06)
This study was designed to examine the interaction of two anti-breast cancer drugs, i.e., fluoxymesterone (FLU) and cyclophosphamide (CYC), with human serum albumin (HSA) using different kinds of spectroscopic, zeta potential and molecular modeling techniques under imitated physiological conditions. The
Alain Berrebi et al.
Haematologica, 92(2), e15-e16 (2007-04-05)
We report a patient with very advanced myelofibrosis and huge splenomegaly who showed a complete hematological response to low dose thalidomide with reversal of splenomegaly and bone narrow fibrosis after 30 months of the treatment.
Oscar J Pozo et al.
Journal of mass spectrometry : JMS, 43(3), 394-408 (2007-11-27)
The suitability of liquid chromatography tandem mass spectrometry (LC-MS/MS) and gas chromatography mass spectrometry (GC-MS) for the elucidation of fluoxymesterone metabolism has been evaluated. Electrospray ionization (ESI) and collision induced dissociation (CID) fragmentation in LC-MS/MS and electron impact spectra (EI)
G Falkson et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 16(5), 1669-1676 (1998-05-20)
To investigate the value of maintenance treatment for patients with metastatic breast cancer whose disease is in complete remission (CR). One hundred ninety-five women (141 eligible) whose disease was in CR or in CR except for bone metastases following six

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service