Skip to Content
Merck
All Photos(1)

Key Documents

F-035

Supelco

Fluconazole-13C3 solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€377.00

€377.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€377.00

About This Item

Empirical Formula (Hill Notation):
C1013C3H12F2N6O
Molecular Weight:
309.25
UNSPSC Code:
41116107
NACRES:
NA.24

€377.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

General description

A stable-labeled internal standard suitable for quantification of fluconazole levels in blood, serum, or plasma by HPLC or LC-MS/MS. Fluconazole, sold under trade names such as Trican, Monicure, Diflucan®, and Afungil, is an antifungal drug used in the treatment and prevention of superficial and systemic fungal infections.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Diflucan is a registered trademark of Pfizer, Inc.
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service