Skip to Content
Merck
All Photos(1)

Key Documents

F-029

Supelco

Felbamate solution

2.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€131.00

€131.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€131.00

About This Item

Empirical Formula (Hill Notation):
C11H14N2O4
CAS Number:
Molecular Weight:
238.24
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€131.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

2.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

NC(=O)OCC(COC(=O)N)c1ccccc1

InChI

1S/C11H14N2O4/c12-10(14)16-6-9(7-17-11(13)15)8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H2,12,14)(H2,13,15)

InChI key

WKGXYQFOCVYPAC-UHFFFAOYSA-N

General description

Felbamate is an antiepileptic drug used for the treatment of epilepsy, partial seizures in adults, and partial and generalized seizures associated with Lennox-Gastautsyndrome in children. The drug has a narrow therapeutic range and carries an increased risk of fatal aplastic anemia and/or liver failure. This certified solution standard is suitable as starting material for calibrators or controls for a variety of LC/MS or GC/MS applications such as forensic analysis, clinical toxicology and therapeutic drug monitorin.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service