Skip to Content
Merck
All Photos(1)

Key Documents

F-024

Supelco

(±)-4-Fluoromethamphetamine-D5 (side chain) hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9D5FN · HCl
Molecular Weight:
208.72
EC Number:
UNSPSC Code:
41116107
Pricing and availability is not currently available.

grade

certified reference material

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

100 μg/mL in methanol (as free base)

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C(C(NC)([2H])C([2H])([2H])[2H])C1=CC=C(F)C=C1.[H]Cl

InChI

1S/C10H14FN.ClH/c1-8(12-2)7-9-3-5-10(11)6-4-9;/h3-6,8,12H,7H2,1-2H3;1H/i1D3,7D,8D;

InChI key

ATENQHYHZHUQLJ-JRXSFSLASA-N

General description

An internal standard suitable for quantitation of 4-Fluoromethamphetamine levels in urine or other matrices by LC/MS or GC/MS for testing applications in forensic analysis, clinical toxicology or urine drug testing. Also known as 4-FMA, 4-Fluoromethamphetamine is sold as a club/designer drug in the illicit market. As a fluoro analog of methamphetamine, the drug produces stimulant and possibly empathogenic or psychoactive effects.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service