Skip to Content
Merck
All Photos(1)

Key Documents

ERI-001

Supelco

Indeno[1,2,3-cd]pyrene

vial of 25 mg, analytical standard, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H12
CAS Number:
Molecular Weight:
276.33
Beilstein:
1879312
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

vial of 25 mg

manufacturer/tradename

Cerilliant®

application(s)

environmental
forensics and toxicology

format

neat

storage temp.

room temp

SMILES string

c1ccc-2c(c1)-c3ccc4ccc5cccc6cc-2c3c4c56

InChI

1S/C22H12/c1-2-7-17-16(6-1)18-11-10-14-9-8-13-4-3-5-15-12-19(17)22(18)21(14)20(13)15/h1-12H

InChI key

SXQBHARYMNFBPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J E Rice et al.
Carcinogenesis, 11(11), 1971-1974 (1990-11-01)
Indeno[1,2,3-cd]pyrene (IP) is a non-alternant polycyclic aromatic hydrocarbon that has tumor-initiating activity on mouse skin and is carcinogenic in newborn mice and in rat lungs. Previous studies have shown that 8- and 9-hydroxyIP and IP-1,2-diol are major metabolites formed in
J E Rice et al.
Cancer research, 45(11 Pt 1), 5421-5425 (1985-11-01)
Indeno[1,2,3-cd]pyrene (IP) is a major environmental pollutant which is carcinogenic on mouse skin and in rat lung. Unlike benzo(a)pyrene, IP is a nonalternant polycyclic aromatic hydrocarbon which is devoid of a bay region. IP was mutagenic in Salmonella typhimurium TA100
J E Rice et al.
Carcinogenesis, 7(10), 1761-1764 (1986-10-01)
Indeno[1,2,3-cd]pyrene is a ubiquitous environmental pollutant which is active as a tumor initiator and complete carcinogen on mouse skin and is carcinogenic in rat lung. The major metabolites of indeno[1,2,3-cd]pyrene as formed in vivo in mouse skin have been identified.
Luke Chandler Short et al.
Journal of the American Society for Mass Spectrometry, 18(4), 589-599 (2006-12-26)
The technique of atmospheric pressure photoionization (APPI) has several advantages over electrospray ionization (ESI) and atmospheric pressure chemical ionization (APCI), including efficient ionization of nonpolar or low charge affinity compounds, reduced susceptibility to ion suppression, high sensitivity, and large linear
E H Pfeiffer et al.
Zentralblatt fur Bakteriologie, Mikrobiologie und Hygiene. 1. Abt. Originale B, Hygiene, 179(6), 574-585 (1984-12-01)
The possible destruction in the soil of PAH present in sewage sludge-garbage composts in considerable amounts has been studied under various conditions. Sandy and loamy soil were mixed with the compost and kept in a climate chamber at 20 degrees

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service