Skip to Content
Merck
All Photos(1)

Key Documents

ERA-018

Supelco

4-Amino-2,6-dinitrotoluene

vial of 100 mg, analytical standard, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7N3O4
CAS Number:
Molecular Weight:
197.15
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

packaging

vial of 100 mg

manufacturer/tradename

Cerilliant®

application(s)

environmental

format

neat

storage temp.

room temp

SMILES string

NC1=CC([N+]([O-])=O)=C(C)C([N+]([O-])=O)=C1

InChI

1S/C7H7N3O4/c1-4-6(9(11)12)2-5(8)3-7(4)10(13)14/h2-3H,8H2,1H3

InChI key

KQRJATLINVYHEZ-UHFFFAOYSA-N

General description

4-Amino-2,6-dinitrotoluene (4A2,6-ADNT) is a metabolite of 2,4,6-trinitrotoluene, which is widely used as an explosive in military shells, bombs, and grenades.

Application

4-Amino-2,6-dinitrotoluene may be used as an analytical standard for the determination of the analyte in explosives residues, ammunition waste, and urine by chromatography techniques.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mehilal et al.
Journal of hazardous materials, 84(2-3), 117-122 (2001-06-19)
A chemoselective reductive method has been achieved for the preparation of 4-picrylamino-2,6-dinitrotoluene (PADNT), a new insensitive energetic material which has been characterised by spectral data and elemental analysis. Some explosive properties of the compound have also been determined and the
B H Woollen et al.
British journal of industrial medicine, 43(7), 465-473 (1986-07-01)
Trinitrotoluene (TNT) absorption was assessed in groups of workers at two explosives factories by measuring the urinary concentrations of dinitroaminotoluene (DNAT) metabolites. DNAT was detected in most of the urine samples analysed and for postshift samples the mean (SD) concentration
G Ahlborg et al.
British journal of industrial medicine, 45(5), 353-358 (1988-05-01)
Urine samples taken after work and after a free weekend from 50 workers employed in various activities in a chemical plant manufacturing explosives were analysed. On the basis of hygienic surveys, the subjects were divided into three categories of exposure
L R Johnson et al.
Ecotoxicology and environmental safety, 27(1), 23-33 (1994-02-01)
Coexposure of 2,4,6-trinitrotoluene (TNT), 2-amino-4,6-dinitrotoluene (2A), or 4-amino-2,6-dinitrotoluene (4A) to near-ultraviolet (nuv) light (lambda max-354 nm) significantly enhanced their toxicity toward Photobacterium phosphoreum (Microtox bioassay) during 30 min but not 15 min. Based on the slopes of the dose-response lines
J Yinon et al.
Toxicology letters, 26(2-3), 205-209 (1985-08-01)
Metabolites of 2,4,6-trinitrotoluene (TNT) were found in the urine of rats fed with TNT. The urine extracts were analysed by liquid chromatography-mass spectrometry (LC-MS). Metabolites found included TNT itself as well as 2-amino-4,6-dinitrotoluene, 4-amino-2,6-dinitrotoluene and 2,4-diamino-6-nitrotoluene, indicating that reduction processes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service