Skip to Content
Merck
All Photos(1)

Key Documents

D-919

Supelco

N-Desmethylflunitrazepam solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10FN3O3
CAS Number:
Molecular Weight:
299.26
EC Number:
UNSPSC Code:
41116107
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

InChI

1S/C15H10FN3O3/c16-10-3-1-2-9(6-10)15-12-7-11(19(21)22)4-5-13(12)18-14(20)8-17-15/h1-7H,8H2,(H,18,20)

InChI key

GAFGVIXGAAPSTO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A certified solution standard suitable for use as starting material in calibrators and controls of N-desmethylflunitrazepam for LC/MS or GC/MS testing methods. N-Desmethylflunitrazepam, also known as norflunitrazepam, is a major urinary metabolite of flunitrazepam. Flunitrazepam is an intermediate-acting benzodiazepine marketed under the trade name Rohypnol as a hypnotic, sedative, anticonvulsant and skeletal muscle relaxant.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service