Skip to Content
Merck
All Photos(1)

Key Documents

D-179

Supelco

Diltiazem-D6 hydrochloride solution

100 μg/mL in acetonitrile ((as free base)), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€480.00

€480.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€480.00

About This Item

Empirical Formula (Hill Notation):
C22H20D6N2O4S · HCl
CAS Number:
Molecular Weight:
457.02
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€480.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile ((as free base))

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

O=C1N(CCN(C([2H])([2H])[2H])C([2H])([2H])[2H])C2=CC=CC=C2S[C@@H](C3=CC=C(OC)C=C3)[C@H]1OC(C)=O.Cl

InChI

1S/C22H26N2O4S.ClH/c1-15(25)28-20-21(16-9-11-17(27-4)12-10-16)29-19-8-6-5-7-18(19)24(22(20)26)14-13-23(2)3;/h5-12,20-21H,13-14H2,1-4H3;1H/t20-,21+;/m1./s1/i2D3,3D3;

InChI key

HDRXZJPWHTXQRI-VEOFFBAHSA-N

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service