Skip to Content
Merck

Skip To

D-103

Desomorphine-D3 solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C17H18D3NO2
CAS Number:
Molecular Weight:
274.37
UNSPSC Code:
41116107
MDL number:
NACRES:
NA.24
EC Number:
200-835-2
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

OC1=C(O2)C([C@@]3([C@]2([H])CCC4)[C@]4([H])[C@H](N(C([2H])([2H])[2H])CC3)C5)=C5C=C1

InChI

1S/C17H21NO2/c1-18-8-7-17-11-3-2-4-14(17)20-16-13(19)6-5-10(15(16)17)9-12(11)18/h5-6,11-12,14,19H,2-4,7-9H2,1H3/t11-,12+,14-,17+/m0/s1/i1D3

InChI key

LNNWVNGFPYWNQE-ACBFFUFASA-N

General description

A certified reference solution and internal standard for quantificatio of desomorphine levels in urine or other matrices by LC/MS or GC/MS. This stable-labeled internal standard is suitable for opiate testing applications in clinical toxicology, forensic analysis, prescription monitoring, or urine drug testing. Desomorphine, most commonly referred to by its street name Krokodil, is a designer opiate and morphine derivative originally abused in Russia and other parts of Europe due to its ease of manufacture from codeine. Desomorphine was previously sold under the trade name Permonid as an opiate drug for treatment of pain.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.


Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

WGK

WGK 2

Flash Point (°F)

35.6 °F - closed cup

Flash Point (°C)

2.0 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters