Skip to Content
Merck
All Photos(1)

Key Documents

D-094

Supelco

N-Desmethylclobazam-8-chloro isomer-13C6 solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€213.00

€213.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€213.00

About This Item

Empirical Formula (Hill Notation):
13C6C9H11ClN2O2
Molecular Weight:
292.67
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€213.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

ClC1=CC(N2[13C]3=[13CH][13CH]=[13CH][13CH]=[13CH]3)=C(NC(CC2=O)=O)C=C1

InChI

1S/C15H11ClN2O2/c16-10-6-7-12-13(8-10)18(11-4-2-1-3-5-11)15(20)9-14(19)17-12/h1-8H,9H2,(H,17,19)/i1+1,2+1,3+1,4+1,5+1,11+1

InChI key

RRTVVRIFVKKTJK-DIYFWHTESA-N

General description

An internal standard for quantification of N-desmethylclobazam levels in urine or other matrices by LC/MS or GC/MS for clinical toxicology, urine drug testing, or forensic analysis applications. Also known as norclobazam, this pharmacologically active biomarker is a major metabolite of clobazam. Clobazam is a benzodiazepine marketed under the brand names Frisium, Urbanol, and Onfi® for use as an anxiolytic and anticonvulsant.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Onfi is a registered trademark of Lundbeck LLC
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2.0 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service