Skip to Content
Merck

Skip To

D-075

N-Desmethyldoxepin-D3 hydrochloride solution

cis/trans, 100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

N-Desmethyldoxepin-D3 hydrochloride solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mL
Check Cart for Availability
€ 170,00

About This Item

Empirical Formula (Hill Notation):
C18D3H16NO · HCl
Molecular Weight:
304.83
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€ 170,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])NCC/C=C1C2=C(C=CC=C2)COC3=C/1C=CC=C3.[H]Cl

InChI

1S/C18H19NO.ClH/c1-19-12-6-10-16-15-8-3-2-7-14(15)13-20-18-11-5-4-9-17(16)18;/h2-5,7-11,19H,6,12-13H2,1H3;1H/b16-10-;/i1D3;

InChI key

GNPPEZGJRSOKRE-XUNJSTFTSA-N

General description

N-Desmethyldoxepin or nordoxepin is the primary metabolite in urine and plasma of doxepin, a tricyclic antidepressant marketed as Apona, Adapine, or Doxal. This internal standard is suitable for numerous GC/MS and LC/MS applications from forensic analysis and clinical toxicology testing to urine drug testing, pain prescription monitoring and isotope dilution methods.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D060E062D074
description

cis/trans, 100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

description

(cis/trans), 100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

description

1.0 mg/mL in methanol (as pyrrolinium), ampule of 1 mL, certified reference material, Cerilliant®

description

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

application(s)

clinical testing

application(s)

clinical testing

application(s)

forensics and toxicology

application(s)

clinical testing

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

form

liquid

form

liquid

form

liquid

form

liquid

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

67-56-1

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters