Skip to Content
Merck

Skip To

C-154

(±)-Cannabicyclol (CBL) solution

1.0 mg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
eCl@ss:
25140904
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

cannabis testing

format

single component solution

storage temp.

−20°C

SMILES string

O1C2(C3C(C(C3c4c1cc(cc4O)CCCCC)(C)C)CC2)C

InChI

1S/C21H30O2/c1-5-6-7-8-13-11-15(22)17-16(12-13)23-21(4)10-9-14-18(21)19(17)20(14,2)3/h11-12,14,18-19,22H,5-10H2,1-4H3

InChI key

IGHTZQUIFGUJTG-UHFFFAOYSA-N

General description

This Certified Spiking Solution® is suitable for use in HPLC, GC/MS or LC-MS/MS CBL testing applications in pharmaceutical research, forensic analysis, or Cannabis potency testing & impurity profiling of Cannabis blends or food products.

(±)-Cannabicyclol is a degradation product of cannabichromene (CBC) formed upon 2 + 2 cyclization of the double bonds in the presence of sunlight or heat.

Application

The Certified Reference Material (CRM) in solution can also be used as follows:
  • Multi-analysis of 11 cannabinoids in biomass and extracts of different cannabis varieties by an HPLC-UV method, following the International Conference on Harmonization (ICH) Tripartite Guideline for Validation of Analytical Procedures
  • Development and validation of an ultra high-performance liquid chromatographic (UHPLC) method coupled to photodiode array and mass spectrometry detectors to determine 11 cannabinoids in 32 Cannabis Sativa samples
  • Separation and estimation of 13 cannabinoids from different cannabis products using isocratic high-performance liquid chromatography (HPLC) in combination with a diode array detector (DAD)
  • Simultaneous determination of 18 cannabinoids and their metabolites from serum samples by LC-MS/MS-based method
  • Quantitative analysis of 17 cannabinoids in cannabis and hemp samples by LC-MS/MS following their liquid-solid extraction in accordance with AOAC and ASTM guidelines for cannabis and hemp matrices

Features and Benefits

  • Fully characterized under ISO/IEC 17025and ISO 17034 accreditation
  • Accompanied with a comprehensive certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

WGK

WGK 2

Flash Point (°F)

35.6 °F - closed cup

Flash Point (°C)

2.0 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters