Skip to Content
Merck
All Photos(1)

Key Documents

C-082

Supelco

Caffeine-13C3 solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Caffeine-trimethyl-13C3 solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€164.00

€164.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€164.00

About This Item

Empirical Formula (Hill Notation):
13C3C5H10N4O2
Molecular Weight:
197.17
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€164.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

[13CH3]N1C(=O)N([13CH3])c2ncn([13CH3])c2C1=O

InChI

1S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3/i1+1,2+1,3+1

InChI key

RYYVLZVUVIJVGH-VMIGTVKRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A stable-labeled internal standard suitable for quantitation of caffeine levels in urine, serum, plasma, or dietary supplements by LC/MS or GC/MS for urine drug testing, clinical toxicology, dietary supplement testing. or isotope dilution methods. Caffeine is an alkaloid compound found naturally in many plants. Caffeine is a central nervous system (CNS) and metabolic stimulant as well as a diuretic.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service