Skip to Content
Merck
All Photos(1)

Key Documents

B-067

Supelco

(±)-Baclofen-D4 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8D4ClNO2
Molecular Weight:
217.69
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

Clc1c(c(c(c(c1[2H])[2H])C(CN)CC(=O)O)[2H])[2H]

InChI

1S/C10H12ClNO2/c11-9-3-1-7(2-4-9)8(6-12)5-10(13)14/h1-4,8H,5-6,12H2,(H,13,14)/i1D,2D,3D,4D

General description

Baclofen, a derivative of gamma-aminobutyric acid (GABA), is a muscle relaxant used to treat spasticity. Marketed with brand names such as Kemstro, Lioresal or Gablofen, the drug has also shown effectiveness in the treatment of alcoholism. This certified solution standard is suitable for use as an internal standard in quantitative mass spectrometry-based testing applications in clinical toxicology, forensic analysis, pharmaceutical research or diagnostic testing.

Application


  • Antidepressant Efficacy in Psychotherapy: Analyzes the effectiveness of combining antidepressants like O-Desmethylvenlafaxine with psychotherapy, enhancing therapeutic strategies for depression treatment (Alemi et al., 2024).

  • Advanced Quantification Methods: Development of a multiplexed liquid chromatography-tandem mass spectrometry method for the quantification of Venlafaxine and its metabolites, including O-Desmethylvenlafaxine, crucial for precise pharmacokinetic studies in clinical settings (Pandey et al., 2024).

  • Pharmacokinetic Modeling: Utilizes physiologically based pharmacokinetic modeling to explore the impact of CYP2D6 and CYP2C19 polymorphisms on Venlafaxine metabolism, crucial for understanding the role of O-Desmethylvenlafaxine in therapeutic efficacy and drug-gene interactions (Shen et al., 2024).

  • Impact of Pharmacogenomics: Studies the influence of CYP2D6 phenotype on the pharmacokinetics of Venlafaxine in elderly patients, highlighting the importance of O-Desmethylvenlafaxine levels in optimizing antidepressant dosing (Men et al., 2024).

  • Environmental Monitoring: Investigates the presence of pharmaceuticals, including O-Desmethylvenlafaxine, in surface waters under the EU Water Framework Directive, emphasizing the need for robust environmental surveillance and pollution control measures (Colzani et al., 2024).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service