Skip to Content
Merck
All Photos(1)

Key Documents

B-020

Supelco

1,4-Butanediol solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10O2
Molecular Weight:
90.12
EC Number:
UNSPSC Code:
41116107
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

OCCCCO

InChI

1S/C4H10O2/c5-3-1-2-4-6/h5-6H,1-4H2

InChI key

WERYXYBDKMZEQL-UHFFFAOYSA-N

General description

An analytical reference standard applicable for use in clinical toxicology, forensic analysis or urine drug testing by LC-MS/MS or GC/MS. 1,4-Butanediol is an industrial solvent which has been abused as a recreational drug for its sedative effect. 1,4-Butanediol is known by the street names "One Comma Four" and "One Four Bee."

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Concepción Domingo et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 15(18), 4001-4005 (2014-10-29)
Arrays of polymer/SWCNT (single-wall carbon nanotube) nanowires supported on a residual nanocomposite film are prepared by melt wetting using porous anodic aluminum oxide (AAO) as a template. The aggregation parameter of SWCNTs extracted from the analysis of their Raman radial
Deivasagayam Dakshinamoorthy et al.
ChemSusChem, 7(10), 2923-2929 (2014-08-21)
The melt polymerization of diglycerol with bicyclic anhydride monomers derived from a naturally occurring monoterpene provides an avenue for polyesters with a high degree of sustainability. The hydrophobic anhydrides are synthesized at ambient temperature via a solvent-free Diels-Alder reaction of
Emilio I Alarcon et al.
Nanoscale, 7(44), 18789-18798 (2015-10-29)
The increasing number of multidrug resistant bacteria has revitalized interest in seeking alternative sources for controlling bacterial infection. Silver nanoparticles (AgNPs), are amongst the most promising candidates due to their wide microbial spectrum of action. In this work, we report
Haoming Pang et al.
Soft matter, 11(34), 6893-6902 (2015-08-04)
In this work we reported a novel graphite doped conductive magnetorheological plastomer (GMRP) with magnetic field dependent electro-conductivity. The conductivity of the GMRPs increased by increasing the content of the graphite particles, while it decreased with the graphite size. When
Dan-Ya Lyu et al.
Journal of chromatography. A, 1393, 1-7 (2015-04-05)
Polymer monolith microextraction (PMME) based on capillary monolithic column is an effective and useful technique to preconcentrate trace analytes from environmental and biological samples. Here, we report the fabrication of a novel aluminum terephthalate metal-organic framework (MIL-53(Al)) incorporated capillary monolithic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service