Skip to Content
Merck
All Photos(1)

Key Documents

BL3H9998ECB5

BLD Pharmatech Co., Ltd.

2,2-Difluoroethyl methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
160.14
MDL number:
Pricing and availability is not currently available.

Iupac Name

2,2-difluoroethyl methanesulfonate

Inchi Code

1S/C3H6F2O3S/c1-9(6,7)8-2-3(4)5/h3H,2H2,1H3

InChI key

JVLBOSKDGOGFEY-UHFFFAOYSA-N

storage temp.

Inert atmosphere,2-8C

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Liquid

Product Link

http://www.bldpharm.com/products/163035-65-2.html

Synonym

2,2-Difluoroethyl methanesulfonate

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

MSDS

https://file.bldpharm.com/static/upload/prosds/bldsds/977/SDS-BD00976403.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library