Skip to Content
Merck
All Photos(1)

Key Documents

LM6001

Avanti

d5-DG ISTD Mix I

Avanti Research - A Croda Brand LM6001

Synonym(s):

d5-DG ISTD Mix I, d5-DG Internal Standard Mixture I, ttoluene:methanol (1:1)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€313.00

€313.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€313.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.25

€313.00


Check Cart for Availability

Request a Bulk Order

form

solution

packaging

pkg of 1 × 1 mL (LM6001-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand LM6001

concentration

~10 μg/mL (Refer to C of A for lot specific concentration. )

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

d5-DG ISTD Mix I has been used as a lipid internal standard in liquid chromatography-tandem mass spectrometry to study the lipid dysregulation in behavioral variant frontotemporal dementia (bvFTD)[1] and as a quantitative lipid standard to monitor different lipid extract concentrations.[2]

Packaging

2 mL Amber Glass Sealed Ampule (LM6001-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipidomics Analysis of Behavioral Variant Frontotemporal Dementia: A Scope for Biomarker Development
Kim WS, et al.
Frontiers in Neurology, 9, 104-104 (2018)
A comprehensive method for lipid profiling by liquid chromatography-ion cyclotron resonance mass spectrometry
Fauland A, et al.
Journal of Lipid Research, 52(12), 2314-2322 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service