Skip to Content
Merck
All Photos(1)

Key Documents

LM4113

Avanti

25-hydroxycholesterol (D6)

Avanti Research - A Croda Brand

Synonym(s):

cholest-5-ene-3β,25-diol(d6)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H40D6O2
CAS Number:
Molecular Weight:
408.69
UNSPSC Code:
12352100
NACRES:
NA.25

form

methanol solution

packaging

pkg of 1 × 1 mL (LM4113-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

~10 μg/mL (Refer to C of A for lot specific concentration. )

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@]12[C@]([C@](CC[C@H](O)C3)(C)C3=CC2)([H])CC[C@@]4(C)[C@@]1([H])CC[C@]4([H])[C@]([H])(C)CCCC(O)(C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C27H46O2/c1-18(7-6-14-25(2,3)29)22-10-11-23-21-9-8-19-17-20(28)12-15-26(19,4)24(21)13-16-27(22,23)5/h8,18,20-24,28-29H,6-7,9-17H2,1-5H3/t18-,20+,21+,22-,23+,24+,26+,27-/m1/s1/i2D3,3D3

InChI key

INBGSXNNRGWLJU-KHKDDIDZSA-N

General description

25-hydroxycholesterol is an oxygenated sterol[1] and a hydroxylated derivative of cholesterol.[2]

Biochem/physiol Actions

25-hydroxycholesterol inhibits cholesterol biosynthesis by downregulating the function of 3-hydroxy-3-methylglutaryl-CoA reductase.[1] It is found to be implicated in atherosclerosis. 25-hydroxycholesterol levels are known to be increased in hypercholesterolemic condition.[2]

Packaging

2 mL Amber Glass Sealed Ampule (LM4113-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher M Adams et al.
The Journal of biological chemistry, 279(50), 52772-52780 (2004-09-29)
The current paper demonstrates that cholesterol and its hydroxylated derivative, 25-hydroxycholesterol (25-HC), inhibit cholesterol synthesis by two different mechanisms, both involving the proteins that control sterol regulatory element-binding proteins (SREBPs), membrane-bound transcription factors that activate genes encoding enzymes of lipid
Cholsoon Jang et al.
Nature medicine, 22(4), 421-426 (2016-03-08)
Epidemiological and experimental data implicate branched-chain amino acids (BCAAs) in the development of insulin resistance, but the mechanisms that underlie this link remain unclear. Insulin resistance in skeletal muscle stems from the excess accumulation of lipid species, a process that
Rosamaria Lappano et al.
PloS one, 6(1), e16631-e16631 (2011-02-10)
The hydroxylated derivatives of cholesterol, such as the oxysterols, play important roles in lipid metabolism. In particular, 25-hydroxycholesterol (25 HC) has been implicated in a variety of metabolic events including cholesterol homeostasis and atherosclerosis. 25 HC is detectable in human
Ken Cheng et al.
Journal of agricultural and food chemistry, 66(33), 8876-8884 (2018-07-26)
Membrane lipids, including sphingolipids and glycerol-phospholipids, are essential in maintaining the skin's barrier function in mammals, but their composition in fish skin and their response to diets have not been evaluated. This study investigated the impacts of reducing dietary eicosapentaenoic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service