Skip to Content
Merck
All Photos(1)

Key Documents

LM1200

Avanti

12:0-13:0 PG

Avanti Research - A Croda Brand

Synonym(s):

1-dodecanoyl-2-tridecanoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€161.00

€161.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€161.00

About This Item

Empirical Formula (Hill Notation):
C31H64NO10P
CAS Number:
Molecular Weight:
641.81
UNSPSC Code:
12352211
NACRES:
NA.25

€161.00


Check Cart for Availability

Request a Bulk Order

form

methanol solution

packaging

pkg of 1 × 1 mL (LM1200-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

~10 μg/mL (Refer to C of A for lot specific concentration.)

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC(O)CO)=O)(OC(CCCCCCCCCCCC)=O)COC(CCCCCCCCCCC)=O.[NH4+]

InChI

1S/C31H61O10P.H3N/c1-3-5-7-9-11-13-15-17-19-21-23-31(35)41-29(27-40-42(36,37)39-25-28(33)24-32)26-38-30(34)22-20-18-16-14-12-10-8-6-4-2;/h28-29,32-33H,3-27H2,1-2H3,(H,36,37);1H3/t28?,29-;/m1./s1

InChI key

ZZSPUZLGIFEDHN-CUZOEGLCSA-N

General description

Phosphatidylglycerol (PG), an anionic phospholipid is a predominant component of the cell membrane.[1] It is localized in the thylakoid membrane of chloroplast in plants.[1] PG is synthesized by phosphatidic acid in E. coli.[2]

Application

12:0-13:0 PG has been used to study the lipid′s interaction with the membrane protein of interest.[3]

Biochem/physiol Actions

Phosphatidylglycerol contributes to photosynthetic activity by maintaining the structural photosynthetic apparatus.[2] It is useful for the biosynthesis of cardiolipin.[1]

Packaging

2 mL Amber Glass Sealed Ampule (LM1200-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct Evidence for Requirement of Phosphatidylglycerol in Photosystem II of Photosynthesis
Hagio M, et al.
Plant Physiology, 39, 407-427 (2010)
Phosphatidylglycerol is Essential for the Development of Thylakoid Membranes in Arabidopsis thaliana
Hagio M, et al.
Nutrition, 43(12), 1456-1464 (2002)
Identifying key membrane protein lipid interactions using mass spectrometry.
Gupta K, et al.
Nature Protocols, 13(5), 1106-1106 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service