Skip to Content
Merck
All Photos(1)

Key Documents

999988C

Avanti

13:0 Diether PC

Avanti Research - A Croda Brand 999988C

Synonym(s):

1,2-di-O-tridecyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H72NO6P
CAS Number:
Molecular Weight:
621.91
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mL (999988C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 999988C

concentration

10 mg/mL (999988C-10mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OCCCCCCCCCCCCC)COCCCCCCCCCCCCC)=O

General description

13:0 Diether PC (phosphocholine) is a class of glycerophospholipids that has two tridecane chains attached to the sn-1 and sn-2 positions of the glycerol backbone by ether bonds. It has choline as the alcohol moiety, attached to the phosphate group.

Application

13:0 Diether PC or 1,2-di-O-tridecyl-sn-glycero-3-phosphocholine is suitable for bicelle preparation and as an internal standard for lipid identification in tissue.

Biochem/physiol Actions

13:0 Diether PC (phosphocholine) is useful for bicelles preparation. Bicelles can be integrated into standard crystallization protocols and in contrast to micelles, bicelles maintain the protein in a more native bilayer environment allowing proteins to be captured in a more biologically relevant orientation.

Packaging

5 mL Clear Glass Sealed Ampule (999988C-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hugo F Azurmendi et al.
Carbohydrate research, 337(10), 905-915 (2002-05-15)
The conformations of two synthetic trisaccharides of blood group A and B (alpha-L-Fucp-(1-->2)-[alpha-D-GalpNAc-(1-->3)]-alpha-D-Galp and alpha-L-Fucp-(1-->2)-[alpha-D-Galp-(1-->3)]-alpha-D-Galp, respectively) and of a type A tetrasaccharide alditol, Fucp-(1-->2)-[alpha-D-GalpNAc-(1-->3)]-beta-D-Galp-(1-->3)-GalNAc-ol, were studied by NMR measurements of one-bond C-H residual dipolar couplings in partially oriented liquid crystal
Caloric restriction and intermittent fasting alter hepatic lipid droplet proteome and diacylglycerol species and prevent diabetes in NZO mice
Baumeier C, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1851(5), 566-576 (2015)
John M Dean et al.
Protein & cell, 9(2), 196-206 (2017-05-20)
Ether lipids, such as plasmalogens, are peroxisome-derived glycerophospholipids in which the hydrocarbon chain at the sn-1 position of the glycerol backbone is attached by an ether bond, as opposed to an ester bond in the more common diacyl phospholipids. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service