Skip to Content
Merck
All Photos(1)

Key Documents

900414P

Avanti

16:0(Alkyne)-18:1 PE

Avanti Research - A Croda Brand, powder

Synonym(s):

1-Hexadec-15-ynoyl-2-oleoyl-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€325.00

€325.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€325.00

About This Item

Empirical Formula (Hill Notation):
C39H72NO8P
CAS Number:
Molecular Weight:
713.96
UNSPSC Code:
12352211
NACRES:
NA.25

€325.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (900414P-1MG)
pkg of 1 × 5 mg (900414P-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

click reagents

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[NH3+])(OC[C@]([H])(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCC#C)=O)=O

InChI key

NJYXOUTYLFAWKP-OTMQOFQLSA-N

Packaging

5 mL Amber Glass Screw Cap Vial (900414P-1MG and 900414P-5MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karine Nozeret et al.
Scientific reports, 9(1), 15978-15978 (2019-11-07)
Lipoprotein modification is an essential process in Gram-negative bacteria. The action of three integral membrane proteins that catalyze the transfer of fatty acids derived from membrane phospholipids or cleave the signal peptide of the lipoprotein substrate result in the formation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service