900127E
Avanti
14(15) EET
Avanti Research™ - A Croda Brand, ethanol solution
Synonym(s):
(±)14(15)-epoxy-5Z,8Z,11Z-eicosatrienoic acid
Select a Size
About This Item
Recommended Products
Assay
99% (TLC)
form
ethanol solution
packaging
pkg of 1 × 500 μg (900127E-500UG)
manufacturer/tradename
Avanti Research™ - A Croda Brand
concentration
500 μg/mL (900127E-500UG)
lipid type
neutral glycerides
shipped in
dry ice
storage temp.
−20°C
SMILES string
O1C(C1C\C=C/C\C=C/C\C=C/CCCC(=O)O)CCCCC
InChI key
JBSCUHKPLGKXKH-ILYOTBPNSA-N
General description
Biochem/physiol Actions
Packaging
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
55.4 °F
Flash Point(C)
13 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service