Skip to Content
Merck
All Photos(1)

Key Documents

900125E

Avanti

17(S)-HDHA

Avanti Research - A Croda Brand 900125E, ethanol solution

Synonym(s):

17(S)-hydroxy Docosahexaenoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€238.00
500 μG
€790.00

€238.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€238.00
500 μG
€790.00

About This Item

Empirical Formula (Hill Notation):
C22H32O3
CAS Number:
Molecular Weight:
344.49
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€238.00


Check Cart for Availability

Request a Bulk Order

form

ethanol solution

packaging

pkg of 1 × 1 mL (900125E-100ug)
pkg of 1 × 1 mL (900125E-500ug)

manufacturer/tradename

Avanti Research - A Croda Brand 900125E

concentration

0.1 mg/mL (900125E-100ug)
0.5 mg/mL (900125E-500ug)

lipid type

neutral lipids
neutral glycerides

shipped in

dry ice

storage temp.

−70°C

SMILES string

CC/C=C\C[C@H](O)/C=C/C=C\C/C=C\C/C=C\C/C=C\CCC(O)=O

InChI

1S/C22H32O3/c1-2-3-15-18-21(23)19-16-13-11-9-7-5-4-6-8-10-12-14-17-20-22(24)25/h3,5-8,11-16,19,21,23H,2,4,9-10,17-18,20H2,1H3,(H,24,25)/b7-5-,8-6-,13-11-,14-12-,15-3-,19-16+/t21-/m0/s1

InChI key

SWTYBBUBEPPYCX-YTQNUIGOSA-N

General description

Docosahexaenoic acid (DHA) is an omega-3 polyunsaturated fatty acid (PUFA) with 22 carbons and six double bonds. The first double bond occurring at position three from the methyl terminus (22:6 n-3).[1] DHA is a component of phospholipid membranes and the myelin sheath. It is the most abundant PUFA found in the brain and localized predominantly to the gray matter.[2]

Biochem/physiol Actions

Docosahexaenoic acid (DHA) exhibits various physiological processes such as regulating signal transduction pathways, neurotransmission, production of neurons and formation of myelin sheath around the axon. It is also associated with the membrane receptor function and synaptic plasticity. DHA supports membrane integrity, fluidity and organization. It is also found to be involved in neuroinflammation. DHA plays a vital role in the development and maturation of the brain.[2] 17(S)-HDHA (hydroxy docosahexaenoic acid) can serve as a precursor for specialized pro-resolving mediators (D-series).[3]

Packaging

2 mL Amber Glass Sealed Ampule (900125E-100ug)
2 mL Amber Glass Sealed Ampule (900125E-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael J Weiser et al.
Nutrients, 8(2), 99-99 (2016-02-24)
Docosahexaenoic acid (DHA) is the predominant omega-3 (n-3) polyunsaturated fatty acid (PUFA) found in the brain and can affect neurological function by modulating signal transduction pathways, neurotransmission, neurogenesis, myelination, membrane receptor function, synaptic plasticity, neuroinflammation, membrane integrity and membrane organization.
Principles of Nutrigenetics and Nutrigenomics: Fundamentals of Individualized Nutrition, 460-460 (2019)
Miaomiao Ruan et al.
Journal of the neurological sciences, 396, 247-253 (2018-12-12)
Glioblastoma (GBM) is a typical malignant tumor, and there are no effective drugs capable of improving patient survival. Docosahexaenoic acid (DHA), a nutrient essential to animal health and neurodevelopment, exerts an anticancer effect in several types of cancer. However, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service