Skip to Content
Merck
All Photos(1)

Key Documents

900108E

Avanti

15β-PGF2α

ethanol solution, Avanti Research - A Croda Brand

Synonym(s):

Prostaglandin F2α (15 beta epimer)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€185.00

€185.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€185.00

About This Item

Empirical Formula (Hill Notation):
C20H34O5
CAS Number:
Molecular Weight:
354.48
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€185.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

ethanol solution

packaging

pkg of 1 × 1 mg

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

250 μg/mL in ethanol

lipid type

neutral lipids
neutral glycerides

shipped in

dry ice

storage temp.

−20°C

Packaging

5 mL Clear Glass Sealed Ampule (900108E-1MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service