Skip to Content
Merck
All Photos(1)

Documents

880122P

Avanti

DSPE-PEG(2000) Cyanur

Avanti Research - A Croda Brand 880122P, powder

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[cyanur(polyethylene glycol)-2000] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (880122P-10mg)
pkg of 1 × 50 mg (880122P-50mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880122P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOO/C=C/NC1=NC(Cl)=NC(Cl)=N1)=O)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C

General description

DSPE-PEG(2000) Cyanur is a conjugate of cationic lipid DSPE-PEG (1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-(polyethylene glycol) and cyanuric groups.

Application

DSPE-PEG has been used in the preparation of cationic liposome. It has also been used to suspend single-walled carbon nanotubes.

Packaging

5 mL Amber Glass Screw Cap Vial (880122P-10mg)
5 mL Amber Glass Screw Cap Vial (880122P-50mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protein-targeted corona phase molecular recognition
Bisker G, et al.
Nature Communications, 7, 10241-10241 (2016)
Targeted delivery of let-7a microRNA encapsulated ephrin-A1 conjugated liposomal nanoparticles inhibit tumor growth in lung cancer
Lee HY, et al.
International journal of nanomedicine, 8, 4481-4481 (2013)
MicroRNA Targeted Cancer Therapy, 110-110 (2014)
Heike Bakowsky et al.
Biochimica et biophysica acta, 1778(1), 242-249 (2007-10-30)
Carbohydrate moieties of the cellular glycocalyx have been suggested to play an important role in biological recognition processes during pathologic conditions, such as inflammation and cancer. Herein, we describe lectin-modified liposomes which might have potential for site-specific drug delivery during
Mahsa Kheradmandi et al.
Cellular and molecular bioengineering, 13(3), 189-199 (2020-05-20)
Atherosclerosis (ATH), the build up of fat in the arteries, is a principal cause of heart attack and stroke. Drug instability and lack of target specificity are major drawbacks of current clinical therapeutics. These undesirable effects can be eliminated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service