Skip to Content
Merck
All Photos(1)

Documents

870718P

Avanti

18:0 Coenzyme A

Avanti Research - A Croda Brand 870718P, powder

Synonym(s):

stearoyl Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H79N10O17P3S
CAS Number:
Molecular Weight:
1085.09
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (870718P-10mg)
pkg of 1 × 5 mg (870718P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870718P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CCCCCCCCCCCCCCCCC)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

InChI

1S/C39H70N7O17P3S.3H3N/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-30(48)67-23-22-41-29(47)20-21-42-37(51)34(50)39(2,3)25-60-66(57,58)63-65(55,56)59-24-28-33(62-64(52,53)54)32(49)38(61-28)46-27-45-31-35(40)43-26-44-36(31)46;;;/h26-28,32-34,38,49-50H,4-25H2,1-3H3,(H,41,47)(H,42,51)(H,55,56)(H,57,58)(H2,40,43,44)(H2,52,53,54);3*1H3/t28-,32?,33+,34+,38-;;;/m1.../s1

InChI key

MQEBLHAOCDTGBF-BMPOWUBJSA-N

General description

18:0 Coenzyme A, also known as stearoyl Coenzyme A, is a coenzyme A derivative of stearic acid.

Application

18:0 Coenzyme A has been used as a constituent of assay buffer in stearoyl-CoA desaturase (SCD1) activity assay.

Packaging

5 mL Amber Glass Screw Cap Vial (870718P-10mg)
5 mL Amber Glass Screw Cap Vial (870718P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haiyan Xu et al.
Frontiers in bioscience : a journal and virtual library, 12, 3781-3794 (2007-05-09)
Stearoyl-CoA desaturases (SCDs) catalyze the biosynthesis of monounsaturated fatty acids from saturated fatty acids. Four scd genes have been identified in mice and three in human (including one pseudogene). Among the four mouse SCD isoforms, SCD1 is predominantly expressed in
Jeong-Won Nam et al.
The Plant cell, 32(4), 820-832 (2020-02-16)
Acyl carrier proteins (ACPs) are the scaffolds for fatty acid biosynthesis in living systems, rendering them essential to a comprehensive understanding of lipid metabolism. However, accurate quantitative methods to assess individual acyl-ACPs do not exist. We developed a robust method

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service