Skip to Content
Merck
All Photos(1)

Key Documents

870713P

Avanti

13:0 Coenzyme A

Avanti Research - A Croda Brand 870713P, powder

Synonym(s):

tridecanoyl Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€373.00

€373.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€373.00

About This Item

Empirical Formula (Hill Notation):
C34H69N10O17P3S
CAS Number:
Molecular Weight:
1014.95
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€373.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (870713P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870713P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CCCCCCCCCCCC)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

General description

13:0 Coenzyme A, also known as tridecanoyl Coenzyme A, is a coenzyme A derivative of tridecanoic acid. 13:0 Coenzyme A is a long-chain acyl CoA.

Application

13:0 Coenzyme A has been used as a spiking standard for mass spectrometry measurements of fatty acid-CoA assay products.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (870713P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Makarova et al.
Current biology : CB, 30(3), 367-380 (2020-01-21)
Membrane function is fundamental to life. Each species explores membrane lipid diversity within a genetically predefined range of possibilities. How membrane lipid composition in turn defines the functional space available for evolution of membrane-centered processes remains largely unknown. We address

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service