Skip to Content
Merck
All Photos(1)

Key Documents

870602P

Avanti

PGPC

Avanti Research - A Croda Brand 870602P, powder

Synonym(s):

1-palmitoyl-2-glutaryl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€123.00
5 MG
€397.00

€123.00


Estimated to ship on17 April 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€123.00
5 MG
€397.00

About This Item

Empirical Formula (Hill Notation):
C29H56NO10P
CAS Number:
Molecular Weight:
609.73
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€123.00


Estimated to ship on17 April 2025


Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (870602P-1mg)
pkg of 1 × 5 mg (870602P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870602P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCC(O)=O)=O)COC(CCCCCCCCCCCCCCC)=O)=O

InChI

1S/C29H56NO10P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-20-28(33)37-24-26(40-29(34)21-18-19-27(31)32)25-39-41(35,36)38-23-22-30(2,3)4/h26H,5-25H2,1-4H3,(H-,31,32,35,36)/t26-/m1/s1

InChI key

CDZVJFRXJAUXPP-AREMUKBSSA-N

General description

1-palmitoyl-2-glutaryl-sn-glycero-3-phosphocholine (PGPC) is a constituent of oxidatively modified low density lipoproteins (oxLDLs).[1] It is synthesized from the non-enzymatic oxidation of a major low-density lipoproteins (LDL) phospholipid, 1-palmitoyl-2-arachidonoyl-sn-glycero-3-phosphocholine (PAPC).[2]

Application

1-palmitoyl-2-glutaryl-sn-glycero-3-phosphocholine (PGPC) has been used to study its effect on plasma membrane nanoplatforms using single-molecule microscopy.[3]

Biochem/physiol Actions

1-palmitoyl-2-glutaryl-sn-glycero-3-phosphocholine (PGPC) acts as an inducer of inflammation, proliferation or apoptosis in cultured vascular smooth muscle cells[1] and macrophages.[2] It is accumulated in atherosclerotic lesions.[1]

Packaging

5 mL Clear Glass Sealed Ampule (870602P-1mg)
5 mL Clear Glass Sealed Ampule (870602P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The oxidized phospholipids POVPC and PGPC inhibit growth and induce apoptosis in vascular smooth muscle cells
Fruhwirth GO, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1761(9), 1060-1069 (2006)
The role of high-density lipoproteins in oxidation and inflammation
Van Lenten B, et al.
Trends in Cardiovascular Medicine, 11(3-4), 155-161 (2001)
Oxidized phospholipids inhibit the formation of cholesterol-dependent plasma membrane nanoplatforms
Brameshuber M, et al.
Biophysical Journal, 110(1), 205-213 (2016)
Alexandra Moumtzi et al.
Journal of lipid research, 48(3), 565-582 (2006-12-01)
Lipid oxidation is now thought to be an initiating and sustaining event in atherogenesis. Oxidatively fragmented phospholipids, namely 1-palmitoyl-2-glutaroyl-sn-glycero-3-phosphocholine (PGPC) and 1-palmitoyl-2-(5-oxovaleroyl)-sn-glycero-3-phosphocholine (POVPC), present in minimally modified LDL and atherosclerotic lesions, have been reported to elicit a wide range of
Marco Di Gioia et al.
Nature immunology, 21(1), 42-53 (2019-11-27)
Pathogen-associated molecular patterns (PAMPs) have the capacity to couple inflammatory gene expression to changes in macrophage metabolism, both of which influence subsequent inflammatory activities. Similar to their microbial counterparts, several self-encoded damage-associated molecular patterns (DAMPs) induce inflammatory gene expression. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service