Skip to Content
Merck
All Photos(1)

Key Documents

870520C

Avanti

18:1 CDP DG

Avanti Research - A Croda Brand 870520C

Synonym(s):

1,2-dioleoyl-sn-glycero-3-(cytidine diphosphate) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H91N5O15P2
CAS Number:
Molecular Weight:
1040.21
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mL (870520C-5mg)
pkg of 1 × 2.5 mL (870520C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870520C

concentration

10 mg/mL (870520C-25mg)
5 mg/mL (870520C-5mg)

lipid type

phosphoglycerides
bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP(OP(OC[C@@H]1[C@@H](O)[C@@H](O)[C@H](N2C(N=C(N)C=C2)=O)O1)([O-])=O)([O-])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[NH4+].[NH4+]

InChI

1S/C48H85N3O15P2.2H3N/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-43(52)61-37-40(64-44(53)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2)38-62-67(57,58)66-68(59,60)63-39-41-45(54)46(55)47(65-41)51-36-35-42(49)50-48(51)56;;/h17-20,35-36,40-41,45-47,54-55H,3-16,21-34,37-39H2,1-2H3,(H,57,58)(H,59,60)(H2,49,50,56);2*1H3/b19-17-,20-18-;;/t40-,41-,45+,46?,47-;;/m1../s1

InChI key

NECNTGAHQCHVGS-BQGZBBTBSA-N

Application

18:1 CDP DG is suitable for liposomes preparation.

Biochem/physiol Actions

18:1 CDP DG or 1,2-dioleoyl-sn-glycero-3-(cytidine diphosphate) produces cytidine 5′?monophosphate by the reaction catalyzed by phosphatidylserine synthase (Pss). CDP-diacylglycerol is derived from diacylglycerol. It serves as a precursor in the production of phosphatidylinositides. Unregulated CDP-diacylglycerol signaling may be associated with the depression.

Packaging

5 mL Clear Glass Sealed Ampule (870520C-25mg)
5 mL Clear Glass Sealed Ampule (870520C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew G Reidenbach et al.
Cell chemical biology, 25(2), 154-165 (2017-12-05)
Human COQ8A (ADCK3) and Saccharomyces cerevisiae Coq8p (collectively COQ8) are UbiB family proteins essential for mitochondrial coenzyme Q (CoQ) biosynthesis. However, the biochemical activity of COQ8 and its direct role in CoQ production remain unclear, in part due to lack
Kimberly R Tyeryar et al.
BMC neuroscience, 9, 12-12 (2008-01-26)
Major depression is a serious mood disorder affecting millions of adults and children worldwide. While the etiopathology of depression remains obscure, antidepressant medications increase synaptic levels of monoamine neurotransmitters in brain regions associated with the disease. Monoamine transmitters activate multiple
Ru-Yin Huang et al.
Current microbiology, 77(5), 710-715 (2020-01-04)
Phosphatidylserine synthase (Pss) is involved in the metabolic pathway in phospholipid synthesis in different organisms. In this study, Pss expression in Vibrio parahaemolyticus was verified through liquid chromatography tandem-mass spectrometry. To analyze the characteristics of Pss, the recombinant Pss was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service