Skip to Content
Merck
All Photos(1)

Key Documents

870437P

Avanti

C17:0 anandamide phosphate

Avanti Research - A Croda Brand 870437P, powder

Synonym(s):

heptadecanoyl ethanolamide phosphate (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€113.00

€113.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€113.00

About This Item

Empirical Formula (Hill Notation):
C19H39NNaO5P
CAS Number:
Molecular Weight:
415.48
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€113.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (870437P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870437P

lipid type

bioactive lipids

shipped in

dry ice

storage temp.

−20°C

General description

C17:0 anandamide phosphate is a phosphate ester of C17:0 anandamide. C17:0 anandamide, also known as heptadecenoylethanolamide, is a N-acyl ethanolamide derivative of heptadecanoic acid.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (870437P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natsuo Ueda et al.
Biochimica et biophysica acta, 1801(12), 1274-1285 (2010-08-26)
Ethanolamides of different long-chain fatty acids constitute a class of endogenous lipid molecules generally called N-acylethanolamines (NAEs). They contain N-arachidonoylethanolamine (anandamide), N-palmitoylethanolamine, and N-oleoylethanolamine, which receive considerable attention because of their actions as an endogenous cannabinoid receptor ligand (endocannabinoid), an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service