Skip to Content
Merck
All Photos(1)

Key Documents

870282C

Avanti

18:1 Biotinyl PE

Avanti Research - A Croda Brand 870282C

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(biotinyl) (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H91N3O10PNaS
CAS Number:
Molecular Weight:
992.31
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

form

liquid

packaging

pkg of 1 × 2.5 mL (870282C-25mg)
pkg of 1 × 4 mL (870282C-100mg)
pkg of 2 × 4 mL (870282C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870282C

concentration

10 mg/mL (870282C-25mg)
25 mg/mL (870282C-100mg)
25 mg/mL (870282C-200mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCN([H])C(CCCC[C@](SC[C@@]1(NC(N2)=O)[H])([C@]12[H])[H])=O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Na+]

InChI

1S/C51H92N3O10PS.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-37-48(56)61-41-44(64-49(57)38-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2)42-63-65(59,60)62-40-39-52-47(55)36-34-33-35-46-50-45(43-66-46)53-51(58)54-50;/h17-20,44-46,50H,3-16,21-43H2,1-2H3,(H,52,55)(H,59,60)(H2,53,54,58);/q;+1/p-1/b19-17-,20-18-;/t44-,45+,46+,50+;/m1./s1

InChI key

ZGFSYDGJPSEMJM-AOAFAMOJSA-M

General description

18:1 Biotinyl PE, also referred as 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(biotinyl), belongs to a class of head group modified functionalized lipids.

Application

18:1 Biotinyl PE is suitable: to tag N-glycolyl-monosialodihexosyl-ganglioside (NGcGM3) in the competitive assay[1], in preparation of small unilamellar vesicle[2], to test the functionalization of Polydimethylsiloxane (PDMS) polymer.[3]

Packaging

5 mL Clear Glass Sealed Ampule (870282C-100mg)
5 mL Clear Glass Sealed Ampule (870282C-200mg)
5 mL Clear Glass Sealed Ampule (870282C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Bo Huang et al.
Lab on a chip, 6(3), 369-373 (2006-03-03)
Polydimethylsiloxane (PDMS) surfaces can be functionalized with biotin groups by adding biotinylated phospholipids to the PDMS prepolymer before curing. The addition of beta-D-dodecyl-N-maltoside (DDM) in the solution blocks non-specific protein binding on these functionalized PDMS surfaces. We characterize the surface
H Zhan
Journal of biochemical and biophysical methods, 41(1), 13-19 (1999-10-08)
A general method for measuring interaction of liposome-protein (or potentially small molecules) was developed. This method utilizes biotinylated liposomes to incubate with interactants. Streptavidin-coated paramagnetic resins were then added and the liposomes (along with bound materials) can be quickly separated
M Virginia Gentilini et al.
Cancer immunology, immunotherapy : CII, 65(5), 551-562 (2016-03-13)
The expression of N-glycolyl-monosialodihexosyl-ganglioside (NGcGM3) in humans is restricted to cancer cells; therefore, it is a tumor antigen. There are measurable quantities of circulating anti-NGcGM3 antibodies (aNGcGM3 Abs) in human serum. Interestingly, some people have circulating Ag-specific immunoglobulins G (IgGs)
Ingrid Albertsson et al.
Scientific reports, 9(1), 17234-17234 (2019-11-23)
Denitrification is a microbial pathway that constitutes an important part of the nitrogen cycle on earth. Denitrifying organisms use nitrate as a terminal electron acceptor and reduce it stepwise to nitrogen gas, a process that produces the toxic nitric oxide
Heun Jin Lee et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(49), 19253-19257 (2008-12-03)
Recent advances have enabled 3-dimensional reconstructions of biological structures in vivo, ranging in size and complexity from single proteins to multicellular structures. In particular, tomography and confocal microscopy have been exploited to capture detailed 3-dimensional conformations of membranes in cellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service