Skip to Content
Merck
All Photos(1)

Key Documents

870242C

Avanti

18:1 Glutaryl PE

Avanti Research - A Croda Brand 870242C

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(glutaryl) (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€151.00

€151.00


Estimated to ship on22 April 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€151.00

About This Item

Empirical Formula (Hill Notation):
C46H83NO11PNa
CAS Number:
Molecular Weight:
880.11
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€151.00


Estimated to ship on22 April 2025


Request a Bulk Order

form

liquid

packaging

pkg of 1 × 2.5 mL (870242C-25mg)
pkg of 2 × 4 mL (870242C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870242C

concentration

10 mg/mL (870242C-25mg)
25 mg/mL (870242C-200mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCN([H])C(CCCC(O)=O)=O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Na+]

InChI

1S/C46H84NO11P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-36-45(51)55-40-42(41-57-59(53,54)56-39-38-47-43(48)34-33-35-44(49)50)58-46(52)37-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h17-20,42H,3-16,21-41H2,1-2H3,(H,47,48)(H,49,50)(H,53,54);/q;+1/p-1/b19-17-,20-18-;/t42-;/m1./s1

InChI key

SUVWNFLYXKTNQV-IEONMYGQSA-M

General description

18:1 Glutaryl phosphoethanolamine, also known as dioleoylphosphatidylethanolamine (DOPE) -glutaryl (GA), is a glutaric acid derivatized DOPE.[1]

Application

18:1 Glutaryl phosphoethanolamine has been used in the preparation of liposomes.[1][2]

Packaging

5 mL Clear Glass Sealed Ampule (870242C-200mg)
5 mL Clear Glass Sealed Ampule (870242C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association and release of prostaglandin E1 from liposomes
Davidson S, et al.
Biochimica et Biophysica Acta - Biomembranes, 1327(1), 97-106 (1997)
Stability of association of 1-O-octadecyl-2-O-methyl-sn-glycero-3-phosphocholine with liposomes is composition dependent
Mayhew E, et al.
Biochimica et Biophysica Acta - Biomembranes, 1329(1), 139-148 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service