Skip to Content
Merck
All Photos(1)

Key Documents

870160C

Avanti

16:0 Ptd Thioethanol

Avanti Research - A Croda Brand 870160C

Synonym(s):

1,2-Dipalmitoyl-sn-Glycero-3-Phosphothioethanol (Sodium Salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€151.00

€151.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€151.00

About This Item

Empirical Formula (Hill Notation):
C37H72O8PNaS
CAS Number:
Molecular Weight:
730.99
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€151.00


Check Cart for Availability

Request a Bulk Order

form

liquid

packaging

pkg of 1 × 2.5 mL (870160C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870160C

concentration

10 mg/mL (870160C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCS)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C37H73O8PS.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-36(38)42-33-35(34-44-46(40,41)43-31-32-47)45-37(39)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h35,47H,3-34H2,1-2H3,(H,40,41);/q;+1/p-1/t35-;/m1./s1

InChI key

AKETWRINAWSUQU-RUQJKXHKSA-M

General description

1,2-Dipalmitoyl-sn-Glycero-3-Phosphothioethanol (DPPTE) or 16:0 Ptd Thioethanol is a thiol modified phospholipid.[1]

Biochem/physiol Actions

1,2-Dipalmitoyl-sn-Glycero-3-Phosphothioethanol (DPPTE) or 16:0 Ptd Thioethanol facilitates the liposome formation by building the lipid layers with the assistance of thiol-anchors at the lipid headgroups.[1]

Packaging

5 mL Clear Glass Sealed Ampule (870160C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of thiolated phospholipids on formation of supported lipid bilayers on gold substrates investigated by surface-sensitive methods
Kilicc, A, et al.
Colloids and Surfaces. B, Biointerfaces, 160, 117-125 (2017)
Peptide-functionalized supported lipid bilayers to construct cell membrane mimicking interfaces
Kilic A and Kok FN
Colloids and Surfaces. B, Biointerfaces, 176, 18-26 (2019)
Lena A Lautscham et al.
Biomaterials, 35(10), 3198-3207 (2014-01-21)
Cell behavior such as cell adhesion, spreading, and contraction critically depends on the elastic properties of the extracellular matrix. It is not known, however, how cells respond to viscoelastic or plastic material properties that more closely resemble the mechanical environment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service