Skip to Content
Merck
All Photos(1)

Key Documents

870110O

Avanti

Triolein (18:1 TG)

Avanti Research - A Croda Brand

Synonym(s):

1,2,3-tri-(9Z-octadecenoyl)-glycerol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€161.00

€161.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€161.00

About This Item

Empirical Formula (Hill Notation):
C57H104O6
CAS Number:
Molecular Weight:
885.43
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€161.00


Check Cart for Availability

Request a Bulk Order

form

liquid

packaging

pkg of 1 × 1 mg (870110O-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

neutral lipids
phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H]C(COC(CCCCCCC/C=C\CCCCCCCC)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI key

PHYFQTYBJUILEZ-IUPFWZBJSA-N

General description

Triolein (18:1 TG) is a nanoemulsion-derived glyceroltrioleate.[1]

Biochem/physiol Actions

Triolein is used in 14C-labeled triolein is used in breath test. This test is a most convenient, sensitive and specific test for the detection of steatorrhea.[2]

Packaging

5 mL Clear Glass Sealed Ampule (870110O-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

626.0 °F - closed cup

Flash Point(C)

330.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stable gas-in-liquid emulsions: production in natural waters and artificial media (2011)
Triolein breath test: a sensitive and specific test for fat malabsorption
Newcomer AD, et al.
Gastroenterology, 76(1), 6-13 (1979)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service