Skip to Content
Merck
All Photos(1)

Key Documents

860833P

Avanti

C6(6-azido) GalCer

Avanti Research - A Croda Brand 860833P, powder

Synonym(s):

D-galactosyl-β-1,1′ N-(6"-azidohexanoyl)-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€289.00

€289.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€289.00

About This Item

Empirical Formula (Hill Notation):
C30H56 N4O8
CAS Number:
Molecular Weight:
600.79
UNSPSC Code:
12352211
NACRES:
NA.25

€289.00


Estimated to ship on28 May 2025


Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (860833P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860833P

lipid type

click reagents

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCN=[N+]=[N-])=O)CO[C@H](O1)[C@H](O)[C@@H](O)[C@H]([C@H]1CO)O

InChI

1S/C30H56N4O8/c1-2-3-4-5-6-7-8-9-10-11-12-13-15-18-24(36)23(33-26(37)19-16-14-17-20-32-34-31)22-41-30-29(40)28(39)27(38)25(21-35)42-30/h15,18,23-25,27-30,35-36,38-40H,2-14,16-17,19-22H2,1H3,(H,33,37)/b18-15+/t23-,24+,25+,27-,28-,29+,30+/m0/s1

InChI key

XAHHWMVRWITDCA-QLCMGMRNSA-N

General description

C6(6-azido) GalCer, also known as D-galactosyl-β-1,1′ N-(6"-azidohexanoyl)-D-erythro-sphingosine is a head group modified lipid comprising an omega-terminal azide. The terminal azide group is involved in a highly specific linking reaction with alkyne-containing reagents, known as ‘click chemistry′, in the presence of a copper (Cu)-containing catalyst. Copper-catalyzed azide−alkyne click chemistry plays a vital role in bioconjugation.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860833P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stanislav I Presolski et al.
Current protocols in chemical biology, 3(4), 153-162 (2011-01-01)
The copper-catalyzed azide-alkyne cycloaddition reaction is widely used for the connection of molecular entities of all sizes. A protocol is provided here for the process with biomolecules. Ascorbate is used as reducing agent to maintain the required cuprous oxidation state.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service