Skip to Content
Merck
All Photos(1)

Key Documents

860825P

Avanti

24:1(2R-OH) Ceramide

Avanti Research - A Croda Brand 860825P, powder

Synonym(s):

N-(2′-(R)-hydroxynervonoyl)-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€289.00

€289.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€289.00

About This Item

Empirical Formula (Hill Notation):
C42H81NO4
CAS Number:
Molecular Weight:
664.10
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€289.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (860825P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860825P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC/C=C/[C@@H](O)[C@@H](NC([C@H](O)CCCCCCCCCCCC/C=C\CCCCCCCC)=O)CO

InChI

1S/C42H81NO4/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-23-25-27-29-31-33-35-37-41(46)42(47)43-39(38-44)40(45)36-34-32-30-28-26-24-16-14-12-10-8-6-4-2/h17-18,34,36,39-41,44-46H,3-16,19-33,35,37-38H2,1-2H3,(H,43,47)/b18-17-,36-34+/t39-,40+,41+/m0/s1

InChI key

AEJPDHWODCHVKB-LDVUWZDOSA-N

General description

Ceramides containing 2-hydroxy fatty acids (hFA) are mainly found in the epidermis.[1] 24:1(2R-OH) Ceramide is a 2R hydroxylated nervonic acid-containing sphingolipid.[2]

Biochem/physiol Actions

Hydroxy fatty acid (hFA)-sphingolipids play a vital role in cell signaling,[3] cell differentiation and apoptosis.[2] These lipids also aid in formation and function of myelin.[4] hFA-ceramide is produced by NAD(P)H-dependent enzyme, fatty acid 2-hydroxylase (FA2H).[5] hFA-ceramides help in epidermal permeability barrier function.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (860825P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inge Zöller et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 28(39), 9741-9754 (2008-09-26)
Sphingolipids containing 2-hydroxylated fatty acids are among the most abundant lipid components of the myelin sheath and therefore are thought to play an important role in formation and function of myelin. To prove this hypothesis, we generated mice lacking a
Helena Maier et al.
The Journal of biological chemistry, 286(29), 25922-25934 (2011-06-02)
2-Hydroxylated fatty acid (HFA)-containing sphingolipids are abundant in mammalian skin and are believed to play a role in the formation of the epidermal barrier. Fatty acid 2-hydroxylase (FA2H), required for the synthesis of 2-hydroxylated sphingolipids in various organs, is highly
Lin Guo et al.
Journal of lipid research, 53(7), 1327-1335 (2012-04-21)
FA 2-hydroxylase (FA2H) is an NAD(P)H-dependent enzyme that initiates FA α oxidation and is also responsible for the biosynthesis of 2-hydroxy FA (2-OH FA)-containing sphingolipids in mammalian cells. The 2-OH FA is chiral due to the asymmetric carbon bearing the
Ana B Herrero et al.
Cancer research, 68(23), 9779-9787 (2008-12-03)
PM02734 is a novel synthetic antitumor drug that is currently in phase I clinical trials. To gain some insight into its mode of action, we used the yeast Saccharomyces cerevisiae as a model system. Treatment of S. cerevisiae with PM02734
Nathan L Alderson et al.
Journal of lipid research, 50(6), 1203-1208 (2009-01-28)
Sphingolipids are ubiquitous components of eukaryotic cells that regulate various cellular functions. In many cell types, a fraction of sphingolipids contain 2-hydroxy fatty acids, produced by fatty acid 2-hydroxylase (FA2H), as the N-acyl chain of ceramide [hydroxyl fatty acid (hFA)-sphingolipids].

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service