Skip to Content
Merck
All Photos(1)

Key Documents

860777C

Avanti

18:0 Dibromo MG

Avanti Research - A Croda Brand 860777C

Synonym(s):

1-(9,10-dibromostearoyl)-rac-glycerol

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€192.00

€192.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€192.00

About This Item

Empirical Formula (Hill Notation):
C21H40Br2O4
CAS Number:
Molecular Weight:
516.35
UNSPSC Code:
12352211
NACRES:
NA.25

€192.00


Estimated to ship on31 May 2025


Request a Bulk Order

form

liquid

packaging

pkg of 1 × 2.5 mL (860777C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860777C

concentration

10 mg/mL (860777C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

OCC([H])(O)COC(CCCCCCCC(Br)C(Br)CCCCCCCC)=O

InChI

1S/C21H40Br2O4/c1-2-3-4-5-7-10-13-19(22)20(23)14-11-8-6-9-12-15-21(26)27-17-18(25)16-24/h18-20,24-25H,2-17H2,1H3

InChI key

XXBCOZLVGAZRSV-UHFFFAOYSA-N

General description

18:0 Dibromo MG, also known as 1-(9,10-dibromostearoyl)-rac-glycerol, is a detergent. It is widely used in lipid mesophase crystallization.

Packaging

5 mL Clear Glass Sealed Ampule (860777C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agnieszka U Blachnio-Zabielska et al.
Rapid communications in mass spectrometry : RCM, 25(15), 2223-2230 (2011-07-08)
Long-chain acyl-coenzymes A (acyl-CoAs) (LCACoA) are the activated forms of long-chain fatty acids and serve as key lipid metabolites. Excess accumulation of intracellular LCACoA, diacylglycerols (DAGs) and ceramides may create insulin resistance with respect to glucose metabolism. We present a
Evgeny V Berdyshev et al.
The Journal of biological chemistry, 284(9), 5467-5477 (2009-01-03)
Novel immunomodulatory molecule FTY720 is a synthetic analog of myriocin, but unlike myriocin FTY720 does not inhibit serine palmitoyltransferase. Although many of the effects of FTY720 are ascribed to its phosphorylation and subsequent sphingosine 1-phosphate (S1P)-like action through S1P(1,3-5) receptors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service