Skip to Content
Merck
All Photos(1)

Key Documents

860760P

Avanti

Azido sphingosine (d14:1)

powder, Avanti Research - A Croda Brand

Synonym(s):

(2S,3R,E)-2-amino-14-azidotetradec-4-ene-1,3-diol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€216.00

€216.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€216.00

About This Item

Empirical Formula (Hill Notation):
C14H28N4O2
CAS Number:
Molecular Weight:
284.40
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€216.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

click reagents

shipped in

dry ice

storage temp.

−20°C

Packaging

5 mL Amber Glass Screw Cap Vial (860760P-1MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service