Skip to Content
Merck
All Photos(1)

Key Documents

860675P

Avanti

Sphinganine-1-Phosphate (d20:0)

Avanti Research - A Croda Brand 860675P, powder

Synonym(s):

D-erythro-sphinganine-1-phosphate (C20 base)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€246.00

€246.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€246.00

About This Item

Empirical Formula (Hill Notation):
C20H44NO5P
CAS Number:
Molecular Weight:
409.54
UNSPSC Code:
12352211
NACRES:
NA.25

€246.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (860675P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860675P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](CCCCCCCCCCCCCCCCC)(O)[C@]([NH3+])([H])COP([O-])(O)=O

InChI

1S/C20H44NO5P/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20(22)19(21)18-26-27(23,24)25/h19-20,22H,2-18,21H2,1H3,(H2,23,24,25)/t19-,20+/m0/s1

InChI key

WEYZCBKZGOIPIU-VQTJNVASSA-N

General description

Sphinganine-1-Phosphate is a phosphorylated derivative of C20 base sphinganine.

Biochem/physiol Actions

Exogenous sphinganine-1-phosphate activates sphingosine-1-phosphate receptor 1 (S1P1). It exhibits protective effects against hepatic and renal ischemia and reperfusion (IR).[1] Sphingosine-1 phosphate and sphinganine-1-phosphate are bioactive lipid mediators. Sphinganine-1-phosphate acts opposite to sphingosine-1 phosphate activities.[2] Increased concentration of sphinganine-1-phosphate acts as a potential biomarker for fumonisin exposure and toxicity in mice.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (860675P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shizhong Bu et al.
Arthritis and rheumatism, 62(7), 2117-2126 (2010-03-24)
Previous studies have revealed a phosphatase and tensin homolog (PTEN)-dependent interaction between the sphingolipid agonist dihydrosphingosine 1-phosphate (dhS1P) and the transforming growth factor beta/Smad3 signaling pathway. This study was undertaken to examine responses of systemic sclerosis (SSc) fibroblasts to sphingosine
Dong-Hyun Kim et al.
Journal of toxicology and environmental health. Part A, 69(23), 2071-2082 (2006-10-25)
Fumonisins are specific inhibitors of ceramide synthase in sphingolipid metabolism. An alteration in sphingolipid metabolism as a result of fumonisin B1 (FB1) exposure is related to cell death, and sphinganine/sphingosine ratio has been used as an indicator of fumonisin exposure
Sang Won Park et al.
Laboratory investigation; a journal of technical methods and pathology, 90(8), 1209-1224 (2010-05-12)
Liver failure due to ischemia and reperfusion (IR) and subsequent acute kidney injury are significant clinical problems. We showed previously that liver IR selectively reduced plasma sphinganine-1-phosphate levels without affecting sphingosine-1-phosphate (S1P) levels. Furthermore, exogenous sphinganine-1-phosphate protected against both liver

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service