Skip to Content
Merck
All Photos(1)

Key Documents

860635P

Avanti

C12 Dihydroceramide (d18:0/12:0)

Avanti Research - A Croda Brand 860635P, powder

Synonym(s):

N-lauroyl-D-erythro-sphinganine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€93.10

€93.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€93.10

About This Item

Empirical Formula (Hill Notation):
C30H61NO3
CAS Number:
Molecular Weight:
483.81
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€93.10


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (860635P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860635P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCCCCCC)=O)[C@]([H])(O)CCCCCCCCCCCCCCC

InChI

1S/C30H61NO3/c1-3-5-7-9-11-13-14-15-16-18-19-21-23-25-29(33)28(27-32)31-30(34)26-24-22-20-17-12-10-8-6-4-2/h28-29,32-33H,3-27H2,1-2H3,(H,31,34)/t28-,29+/m0/s1

InChI key

UHWYQXNZIBLESO-URLMMPGGSA-N

Application

C12 Dihydroceramide has been used as an internal standard for quantification of dihydroceramides in adipose tissue by liquid chromatography - tandem mass spectrometry (LC-MS-MS).[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860635P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

George D Mcilroy et al.
Biochemical pharmacology, 100, 86-97 (2015-11-26)
Fenretinide (FEN) is a synthetic retinoid that inhibits obesity and insulin resistance in high-fat diet (HFD)-fed mice and completely prevents 3T3-L1 pre-adipocyte differentiation. The aim of this study was to determine the mechanism(s) of FEN action in 3T3-L1 adipocytes and
Fenretinide mediated retinoic acid receptor signalling and inhibition of ceramide biosynthesis regulates adipogenesis, lipid accumulation, mitochondrial function and nutrient stress signalling in adipocytes and adipose tissue
Mcilroy GD, et al.
Biochemical Pharmacology, 100, 86-97 (2016)
Gergely Karsai et al.
The Journal of biological chemistry, 295(7), 1889-1897 (2019-12-22)
Sphingolipids (SLs) are structurally diverse lipids that are defined by the presence of a long-chain base (LCB) backbone. Typically, LCBs contain a single Δ4E double bond (DB) (mostly d18:1), whereas the dienic LCB sphingadienine (d18:2) contains a second DB at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service