Skip to Content
Merck
All Photos(1)

Key Documents

860608P

Avanti

N-02:0 Phytosphingosine

Avanti Research - A Croda Brand 860608P, powder

Synonym(s):

N-acetoyl 4-hydroxysphinganine (Saccharomyces Cerevisiae)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€313.00

€313.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€313.00

About This Item

Empirical Formula (Hill Notation):
C20H41NO4
CAS Number:
Molecular Weight:
359.54
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€313.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (860608P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860608P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@]([H])(CCCCCCCCCCCCCC)[C@](O)([H])[C@@]([H])(NC(C)=O)CO

InChI

1S/C20H41NO4/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-19(24)20(25)18(16-22)21-17(2)23/h18-20,22,24-25H,3-16H2,1-2H3,(H,21,23)

InChI key

SZUJJDLBXJCDNT-UHFFFAOYSA-N

General description

Phytosphingosine is a sphingoid base with hydroxyl group at the C4 position.[1] It is highly found in plants and fungi. In addition, phytosphingosine is also present in mammalian epidermis, including the stratum corneum.[2]

Biochem/physiol Actions

Phytosphingosine acts as a sphingoid backbone for phytoceramide, which is a yeast analogue of the mammalian ceramide.[1] Phytosphingosine and its derivatives are allied with body′s natural defense system. These lipids prevent dehydration of skin, control epidermal cell growth, differentiation, and apoptosis. In addition, they also exhibit anti-bactericidal and anti-inflammatory activities. Thus, Phytosphingosine and its derivatives is essential for maintaining healthy skin. But, at high concentrations these compounds are toxic.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (860608P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ceramide function in the brain: when a slight tilt is enough
Mencarelli C and Martinez-Martinez, P
Cellular and Molecular Life Sciences, 70(2), 181-203 (2013)
Byung-Hak Kim et al.
The Journal of investigative dermatology, 134(4), 1023-1032 (2013-11-02)
Phytosphingosine is abundant in plants and fungi and is found in mammalian epidermis, including the stratum corneum. Phytosphingosine and its derivatives N-acetyl phytosphingosine and tetraacetyl phytosphingosine are part of the natural defense system of the body. However, these molecules exhibit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service